Vinylacetat 99+% stabilised
Leverantör: Thermo Scientific
Fara
Synonyms: Acetic acid vinyl ester
140840010 140840025 140840250 140840100
ACRO140840010EA 650 SEK
ACRO140840010 ACRO140840025 ACRO140840250 ACRO140840100
Vinylacetat 99+% stabilised
Vinylacetat
Formel: C₄H₆O₂
MW: 86,09 g/mol
Kokpunkt: 72…73 °C (760 mmHg)
Smältpunkt: –93 °C
Densitet: 0,93 g/cm³
Flampunkt: –8 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008713
CAS nummer: 108-05-4
EINECS: 203-545-4
UN: 1301
ADR: 3,II
Merck Index: 12,10130
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3940 to 1.3960 (20°C, 589 nm)
Water ≤0.05 %
GC ≥99.0 %
Stabilizer 3 to 15 ppm Hydroquinone
Acidity (CH3COOH) ≤0.005 %