Erbium(III) perchlorate hexahydrate 50% w/w aqueous solution, reagent
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Erbium perchlorate hexahydrate
40576.NA 40576.NC 40576.NE
40576.NAEA 1150 SEK
40576.NA 40576.NC 40576.NE
Erbium(III) perchlorate hexahydrate 50% w/w aqueous solution, reagent
Erbium(III) perchlorate hexahydrate
CAS No.: 14692-15-0 Molecular Formula: Er(ClO4)3.6H2O Formula Weight: 573.70 (465.61anhy) Physical Form: Liquid Appearance: Pink Odour: Odourless UN No.: 3211 MDL No.: MFCD00150232
Formel: Cl₃ErO₁₂·6H₂O
MW: 573.7
Storage Temperature: Omgivande
MDL Number: MFCD00150232
CAS nummer: 14692-15-0
EINECS: 237-840-4
UN: 3211
ADR: 5.1 II