Hyamine® 1622 0.004 mol/l, Titripur® volumetrisk lösning, for the determination of anionic tensides, Supelco®
Leverantör: Merck
M-Clarity™ quality level = MQ100
1.15480.1000 1.15480.1003
1.15480.1000EA 645 SEK
1.15480.1000 1.15480.1003
Hyamine® 1622 0.004 mol/l, Titripur® volumetrisk lösning, for the determination of anionic tensides, Supelco®
Bensatoniumklorid
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formel: C₂₇H₄₂ClNO₂
Densitet: 1,0 g/cm3 (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011742
CAS nummer: 121-54-0
EINECS: 204-479-9