Decanoyl chloride ≥98%

Leverantör: Thermo Scientific
Fara

Synonyms: Decanoylchloride

A19486.22 A19486.36
A19486.22EA 880 SEK
A19486.22 A19486.36
Decanoyl chloride ≥98%
Decanoyl chloride

Purity: 98% CAS No.: 112-13-0 Molecular Formula: CH3(CH2)8COCl Formula Weight: 190.71 Flash Point: 106°(223°F) Refractive Index: 1.4410 Odour: Irritating Melting Point: -35° Boiling Point: 232° Beilstein Reference: 507055 UN No.: 3265 MDL No.: MFCD00000771

Formel: C₁₀H₁₉ClO
MW: 190,71 g/mol
Kokpunkt: 232 °C
Smältpunkt: –35 °C
Densitet: 0,925
Flampunkt: 106 °C (223 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000771
CAS nummer: 112-13-0
EINECS: 203-938-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR