Decanoyl chloride 98+%

Leverantör: Thermo Scientific
Fara

Synonyms: Decanoylchloride

161191000 161195000
ACRO161191000EA 745 SEK
ACRO161191000 ACRO161195000
Decanoyl chloride 98+%
Decanoyl chloride
Formel: C₁₀H₁₉ClO
MW: 190,71 g/mol
Kokpunkt: 233 °C (1013 hPa)
Smältpunkt: –34,5 °C
Densitet: 0,919 g/cm³
Flampunkt: 110 °C
MDL Number: MFCD00000771
CAS nummer: 112-13-0
EINECS: 203-938-0
UN: 3265
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4430 (20°C, 589 nm)
GC ≥98.0 %
Color scale ≤50 APHA
Carboxylic acid ≤2 % (as Decanoic acid)
Titration RCOCl ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR