3-Quinolinecarboxylic acid

Leverantör: Molekula

65676498-1G
MOLE65676498-1GEA 0 SEK
MOLE65676498-1G
3-Quinolinecarboxylic acid
3-Quinolinecarboxylic acid
Formel: C₁₀H₇NO₂
MW: 173,17 g/mol
Kokpunkt: 345 °C (1013 hPa)
Smältpunkt: 282…283 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006770
CAS nummer: 6480-68-8
EINECS: 229-337-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR