Bromkresolgrönt natriumsalt ACS
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate
402940050 402940100 402940500
ACRO402940050EA 930 SEK
ACRO402940050 ACRO402940100 ACRO402940500
Bromkresolgrönt natriumsalt ACS
Bromkresolgrönt natriumsalt
Formel: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
Smältpunkt: 230 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00148898
CAS nummer: 67763-24-0
EINECS: 263-657-4
Order Now
Specifikation testresultat
Appearance Dark brown to green-black Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.8 (yellow) to pH 5.4 (blue)
Clarity of solution Passes test