4-Methoxystyrene ≥98% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Methoxy-4-vinylbenzene, 4-Vinylanisole
Stabilization: med 4-tert-Butylpyrokatekol
B22353.06 B22353.14 B22353.22
B22353.06EA 1625 SEK
B22353.06 B22353.14 B22353.22
4-Methoxystyrene ≥98% stabilised
4-Methoxystyrene
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 98% CAS No.: 637-69-4 Molecular Formula: C9H10O Formula Weight: 134.18 Storage Temperature: 2°C to 8°C Flash Point: 72°(161°F) Refractive Index: 1.5610 Boiling Point: 204-205° Beilstein Reference: 1098935 MDL No.: MFCD00008619
Formel: C₉H₁₀O
MW: 134,18 g/mol
Kokpunkt: 204…205 °C
Smältpunkt: 3,3 °C
Densitet: 1,002
Flampunkt: 72 °C (161 °F)
MDL Number: MFCD00008619
CAS nummer: 637-69-4
EINECS: 211-298-9