Etylendi(acetat) ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethane-1,2-diyl diacetate, Ethylene glycol diacetate, Ethylene Glycol Diacetate, 1,2-Diacetoxyethane
41910.22 41910.36 41910.0E
41910.22EA 655 SEK
41910.22 41910.36 41910.0E
Etylendi(acetat) ≥97%
Etylendi(acetat)
Formel: C₆H₁₀O₄
MW: 146,14 g/mol
Kokpunkt: 186…187 °C
Smältpunkt: –41 °C
Densitet: 1,13
Flampunkt: 82 °C (181 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008718
CAS nummer: 111-55-7
EINECS: 203-881-1
Merck Index: 14,3799