Kolkicin 95%
Leverantör: Thermo Scientific
Fara
Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine
A13240.03 A13240.06 A13240.14
A13240.03EA 2025 SEK
A13240.03 A13240.06 A13240.14
Kolkicin 95%
Kolkicin
Purity: 95% CAS No.: 64-86-8 Molecular Formula: C22H25NO6 Formula Weight: 399.44 Melting Point: ca 150° dec. Beilstein Reference: 2228813 Merck Reference: 14,2471 UN No.: 1544 MDL No.: MFCD00078484
Formel: C₂₂H₂₅NO₆
MW: 399,44 g/mol
Smältpunkt: ∼ 150 °C (dec.)
Densitet: 1,32 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00078484
CAS nummer: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1,I
Merck Index: 14,2471