Styren-D8 98+ atom% D Stabilised NMR
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Vinylbenzen-D8
175390050 175390010
ACRO175390050EA 6850 SEK
ACRO175390050 ACRO175390010
Styren-D8 98+ atom% D Stabilised NMR
Styren-D8
Formel: C₆D₅CDCD₂
MW: 112.09 g/mol
Kokpunkt: 50…51 °C (25 mmHg)
Smältpunkt: –31 °C
Densitet: 0.97 g/cm³
Flampunkt: 32 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00044231
CAS nummer: 19361-62-7
EINECS: 242-995-6
UN: 2055
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
Stabilizer 0.5 % 4-tert-Butylcatechol
D-Enrichment ≥98.0 %