Pregnenolone-16-α-carbonitrile ≥99%
Leverantör: ENZO LIFE SCIENCES
BMLS6200020 BMLS6200100
ENZOBMLS6200020EA 710 SEK
ENZOBMLS6200020 ENZOBMLS6200100
Pregnenolone-16-α-carbonitrile ≥99%
Pregnenolone-16-alpha-carbonitrile
Cytochrome P450 inducer.
Activates transcription of the PCN1 gene. SXR/PXR Ligand.
Soluble in DMF (5 mg/ml) or acetone (1 mg/ml).
Formel: C₂₂H₃₁NO₂
MW: 341.5 g/mol
Storage Temperature: Frys
CAS nummer: 1434-54-4
Order Now
Specifikation testresultat
Purity ≥99% (HPLC)