Saltsyra 0.1 M
Leverantör: ENZO LIFE SCIENCES
Synonyms: Hydrochloric acid solution, Muriatic acid
ADI800080 ADI800082
ENZOADI800080EA 820 SEK
ENZOADI800080 ENZOADI800082
Saltsyra 0.1 M
Saltsyra
Assay diluent and lysis reagent for cyclic nucleotides.
Formel: HCl
MW: 36.46 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0