Daptomycin ≥93%

Leverantör: ENZO LIFE SCIENCES

BMLA2010020 BMLA2010100
ENZOBMLA2010020EA 1275 SEK
ENZOBMLA2010020 ENZOBMLA2010100
Daptomycin ≥93%
Daptomycin

Lipopeptide antibiotic


Novel lipopeptide antibiotic active against Gram positive bacteria.

Formel: C₇₂H₁₀₁N₁₇O₂₆
MDL Number: MFCD08282794
CAS nummer: 103060-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR