Aflatoxin G1 ≥98%

Leverantör: ENZO LIFE SCIENCES
Fara

ALX630104M001 ALX630104M005
ENZOALX630104M001EA 1800 SEK
ENZOALX630104M001 ENZOALX630104M005
Aflatoxin G1 ≥98%
Aflatoxin G1

Potent hepatotoxic and hepatocarcinogenic mycotoxin.


Soluble in DMSO (10 mg/ml), methylene chloride (5 mg/ml) or methanol.

Formel: C₁₇H₁₂O₇
MW: 328.3 g/mol
Storage Temperature: Kylskåp
CAS nummer: 1165-39-5
EINECS: 214-615-9
UN: 2811
ADR: 6.1 I
Merck Index: 14,00181

Order Now

Specifikation testresultat

Source Isolated from Aspergillus flavus
Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR