Ochratoxin A ≥98% (by HPLC, TLC)
Leverantör: ENZO LIFE SCIENCES
Fara
ALX630089M001 ALX630089M005 ALX630089M025
ENZOALX630089M001EA 745 SEK
ENZOALX630089M001 ENZOALX630089M005 ENZOALX630089M025
Ochratoxin A ≥98% (by HPLC, TLC)
Ochratoxin A
Mycotoxin. Nephrotoxic.
Mycotoxin. Natural contaminant of mouldy food and feed. It has a number of toxic effects, the most prominent being nephrotoxicity. Immunosuppressive, genotoxic, teratogenic and carcinogenic. Stimulates lipid peroxidation. Induces oxidative DNA lesions coupled with direct DNA adducts via quinone formation.
Isolated from Aspergillus ochraceus.
Soluble in methanol (10 mg/ml).
Formel: C₂₀H₁₈ClNO₆
MW: 403.8 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00078079
CAS nummer: 303-47-9
UN: 2811
ADR: 6.1 II
Merck Index: 14,06740
Order Now
Specifikation testresultat
Purity ≥98% (HPLC, TLC)