Isopropyl-β-D-thiogalactopyranoside 98 HPLC_ANALYSMETOD
Leverantör: ENZO LIFE SCIENCES
Varning
Synonyms: IPTG
ALX582012G005 ALX582012G025
ENZOALX582012G005EA 1725 SEK
ENZOALX582012G005 ENZOALX582012G025
Isopropyl-β-D-thiogalactopyranoside 98 HPLC_ANALYSMETOD
Isopropyl-β-D-thiogalactopyranoside
β-galactosidase activator.
Inducer of β-D-galactosidase. Used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function. Contains no animal-derived products.
Soluble in water (5%) or methanol.
Formel: C₉H₁₈O₅S
MW: 238.31 g/mol
Kokpunkt: 402
Storage Temperature: Frys
MDL Number: MFCD00063273
CAS nummer: 367-93-1
EINECS: 206-703-0
Order Now
Specifikation testresultat
Purity ≥98% (HPLC)
Identity Determined by IR