Trifluoperazine dihydrochloride ≥98%
Leverantör: ENZO LIFE SCIENCES
Synonyms:
10-[3-(4-Methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine Dihydrochloride
ALX550310G005
ENZOALX550310G005EA
1900
SEK
ENZOALX550310G005
Trifluoperazine dihydrochloride ≥98%
Trifluoperazine dihydrochloride
Antipsychotic
Facilitates agonist induced elevation of cytosolic calcium at low concentrations (10µM). Antagonizes calmodulin at higher concentrations. Antipsychotic and sedative dopamine antagonist. Potent and irreversible inhibitor of cAMP-gated cationic channels. Inhibits DNA repair. Cell permeable. Potent neurotoxin.
Formel:
C₂₁H₂₄F₃N₃S MW: 480.42 g/mol |
MDL Number:
MFCD00012656 CAS nummer: 440-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...