Histamine dihydrochloride

Leverantör: ENZO LIFE SCIENCES

Synonyms: Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride

ALX550132G005
ENZOALX550132G005EA 600 SEK
ENZOALX550132G005
Histamine dihydrochloride
Histamine dihydrochloride
Formel: C₅H₁₁Cl₂N₃
MW: 184.07 g/mol
MDL Number: MFCD00012703
CAS nummer: 56-92-8
EINECS: 200-298-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR