MnTMPyP pentachloride ≥95%, black powder
Leverantör: ENZO LIFE SCIENCES
SOD mimetic. Peroxynitrite scavenger.
Cell permeable superoxide dismutase (SOD) mimetic and peroxynitrite scavenger. Catalyzes the dismutation of O²⁻ even in the presence of excess EDTA. Displays a protective effect against H₂O₂ mediated injury in astroglial cells.
Soluble in water or aqueous buffers.
Formel:
C₄₄H₃₆MnN₈Cl₅ Storage Temperature: Frys |
CAS nummer:
100012-18-8 |
Specifikation testresultat
Purity (TLC) | ≥95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...