Apoptosis activator 2 ≥98%

Leverantör: ENZO LIFE SCIENCES

Synonyms: 1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione, 1-(3,4-Dichlorobenzyl)indoline-2,3-dione

ALX420035M005 ALX420035M025
ENZOALX420035M005EA 550 SEK
ENZOALX420035M005 ENZOALX420035M025
Apoptosis activator 2 ≥98%
Apoptosis activator 2

Apoptosis inducer.


Induces apoptosis by promoting the oligomerisation of Apaf-1 into the mature, functional apoptosome, triggering caspase-9 activation resulting in activated caspase-3. Shows strong selectivity for cancer cells against normal cell lines. Acts in a similar way like apoptosis inducer PETCM.


Soluble in DMSO (5 mg/ml), methanol (5 mg/ml) or 100% ethanol.

Formel: C₁₅H₉Cl₂NO₂
MW: 306.142 g/mol
Smältpunkt: 174…175 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00141556
CAS nummer: 79183-19-0

Order Now

Specifikation testresultat

Purity ≥98% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR