Ponasterone A ≥95%

Leverantör: ENZO LIFE SCIENCES

ALX370014M001 ALX370014M005
ENZOALX370014M001EA 930 SEK
ENZOALX370014M001 ENZOALX370014M005
Ponasterone A ≥95%
Ponasterone A

Ecdysteroid.


A member of the ecdysteroid family. Analog of ecdysone with similar properties to muristerone A. It is a functional, reliable and economical substitute for muristerone A as an inducer for the ecdysone-inducible mammalian expression system. Results show that ponasterone A was able to induce expression of β-galactosidase to levels similar to those obtained with muristerone A induction. Potent compound for ecdysteroid activity; EC₅₀ = 0.07 µM.


Soluble in acetic acid, 100% ethanol, methanol or DMSO; insoluble in water.

Formel: C₂₇H₄₄O₆
Storage Temperature: Frys
CAS nummer: 13408-56-5
Merck Index: 14,07588

Order Now

Specifikation testresultat

Purity ≥95% (HPLC)
Identity Identity determined by ¹H- and ¹³C-NMR
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR