ABT-888 ≥98%

Leverantör: ENZO LIFE SCIENCES

Synonyms: (R)-2-(2-Methylpyrrolidin-2-yl)-1H-benzo[d]imidazole-7-carboxamide dihydrochloride, Veliparib 2HCl

ALX270444M001 ALX270444M005 ALX270444M025
ENZOALX270444M001EA 1350 SEK
ENZOALX270444M001 ENZOALX270444M005 ENZOALX270444M025
ABT-888 ≥98%
ABT-888

PARP inhibitor


Potent inhibitor of PARP-1 and PARP-2 (potency ≤5nM in vitro). Does not inhibit other NAD-binding enzymes. Has minimal CYP450 inhibition and induction. Shows broad spectrum of chemo- and radiopotentiation. Is toxic to both oxic and hypoxic cells. Enantiomeric purity ≥97% suitable for in vivo studies. Does not show inherent cytotoxicity and shows no single agent activity in tumor models. Has excellent bioavailability and good blood-brain permeation. Increases tumor growth delay resulting from radiation and DNA-damaging agents.

Formel: C₁₃H₁₈Cl₂N₄O
MW: 317.214 g/mol
MDL Number: MFCD12407402
CAS nummer: 912445-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR