Sirtinol ≥97%

Leverantör: ENZO LIFE SCIENCES

ALX270308M001 ALX270308M005
ENZOALX270308M001EA 575 SEK
ENZOALX270308M001 ENZOALX270308M005
Sirtinol ≥97%
Sirtinol

SIRT inhibitor


Specific cell permeable inhibitor of the sirtuin family of NAD-dependent deacetylases (ySir2: IC50=48µM; hSIRT1: IC50=60µM; hSIRT2: IC50=58µM) with no effect on human HDAC1. Reported to inhibit Sir2p transcriptional silencing activity in vivo (IC50=25µM) and NAD-dependent histone deacetylase activity of purified recombinant yeast Sir2p (IC50=70µM) and hSIRT2 (IC50=40µM) in vitro.

Formel: C₂₆H₂₂N₂O₂
MDL Number: MFCD00810186
CAS nummer: 410536-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR