Rosmarinic acid ≥98%

Leverantör: ENZO LIFE SCIENCES

ALX270253M010 ALX270253M050
ENZOALX270253M010EA 550 SEK
ENZOALX270253M010 ENZOALX270253M050
Rosmarinic acid ≥98%
Rosmarinic acid

Naturally occurring polyphenolic compound with antioxidant and anti-inflammatory properties.Inhibits several complement-dependent inflammatory processes via inhibition of the C5 convertase. Anticarcinogenic. Inhibitor of lipid peroxidation, TCR-induced T cell activation and proliferation.


Isolated from Rosmarinus officinalis.

Formel: C₁₈H₁₆O₈
MDL Number: MFCD00017740
CAS nummer: 20283-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR