AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Leverantör: ENZO LIFE SCIENCES
Varning
Synonyms: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
ALX270022G001 ALX270022M250
ENZOALX270022G001EA 7650 SEK
ENZOALX270022G001 ENZOALX270022M250
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Formel: C₈H₁₁ClFNO₂S
MW: 239,69 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00132962
CAS nummer: 30827-99-7
UN: 3261
ADR: 8,II