SB590885 dihydrochloride ≥98%
Leverantör: ENZO LIFE SCIENCES
ENZCHM1130005
ENZCHM1130010
ENZOENZCHM1130005EA
2000
SEK
ENZOENZCHM1130005
ENZOENZCHM1130010
SB590885 dihydrochloride ≥98%
SB590885 dihydrochloride
Potent B-Raf inhibitor.
Selective for B-Raf against 46 other kinases. SB 590885 dihydrochloride decreases anchorage-independent growth of melanoma cell lines. Inhibits ERK phosphorylation and proliferation in tumor cells expressing B-RafV600E.
Formel:
C₂₇H₂₇N₅O₂·2HCl MW: 539 g/mol |
CAS nummer:
405554-55-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...