AdipoRon ≥98%
ENZCHM1010005
ENZCHM1010010
ENZOENZCHM1010005EA
1125
SEK
ENZOENZCHM1010005
ENZOENZCHM1010010
AdipoRon ≥98%
AdipoRon
Novel and orally active adiponectin receptor agonist for AdipoR1 and AdipoR2.
AdipoR agonist that binds to both AdipoR1 and AdipoR2.
Formel:
C₂₇H₂₈N₂O₃ |
MDL Number:
MFCD08271060 CAS nummer: 924416-43-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...