7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid ≥98%
Leverantör: ENZO LIFE SCIENCES
Synonyms:
7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid
BMLEI3890005
BMLEI3890025
ENZOBMLEI3890005EA
1275
SEK
ENZOBMLEI3890005
ENZOBMLEI3890025
7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid ≥98%
STO-609
CaM kinase kinase inhibitor
STO-609 is a specific CaM kinase kinase (CaM-KK) inhibitor. Ki=0.2µM and 0.04µM for alpha and beta isotypes respectively. Demonstrates >80-fold selectivity for CAM-KK over CaMK1, CaMK2, CaMK4, MLCK, PKA, PKC, and p42 MAPK. Concentrations up to 10µg/ml do not affect cellular viability. It is a tool for exploring the role of the CaM-KK pathway in various physiological processes.
Formel:
C₁₉H₁₀N₂O₃ MW: 314.29 g/mol |
MDL Number:
MFCD30685721 CAS nummer: 52029-86-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...