Digoxigenin NHS ester ≥80%
Leverantör: ENZO LIFE SCIENCES
ENZ450220001 ENZ450220005
ENZOENZ450220001EA 3050 SEK
ENZOENZ450220001 ENZOENZ450220005
Digoxigenin NHS ester ≥80%
Digoxigenin NHS ester
Digoxigenin NHS ester is a non-radioactive immune-tag, widely used for in situ hybridisation and protein labeling.
  • Detects interested biomolecules with high affinity and specificity
  • Reacts with amino groups within biomolecules (nucleic acid, proteins, etc) and enables the attachment of digoxigenin into these molecules
Formel: C₃₅H₅₀N₂O₁₀
MW: 658.79 g/mol
Storage Temperature: Frys
CAS nummer: 129273-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da7e806a255287bd5735ea193c64a4847454c08a&vid=30ff9897a62482819e3d5a65c8707e84f5802ec4&prid=web&clid=SAPDAM