Swainsonine 98 HPLC_ANALYSMETOD
Leverantör: ENZO LIFE SCIENCES
Synonyms: 8α,β-Octahydroindolizidine-1α,2α,8β-triol
BMLS1120001 BMLS1120005
ENZOBMLS1120001EA 2350 SEK
ENZOBMLS1120001 ENZOBMLS1120005
Swainsonine 98 HPLC_ANALYSMETOD
Swainsonine
Reversibly inhibits the active site of lysosomal α-mannosidase. Also inhibits the growth of tumors and prevents metastasis in murine models. Could inhibit cell proliferation in vitro and the growth of human gastric carcinoma in vivo. Blocks the processing of high mannose oligosaccharides to form complex type oligosaccharides.^Immunomodulator.
  • Lysosomal alpha-mannosidase inhibitor
White to off-white solid.
Soluble in 100% ethanol, DMSO, methanol or water (10mg/ml).
Formel: C₈H₁₅NO₃
MW: 173.2 g/mol
Storage Temperature: Kylskåp
CAS nummer: 72741-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26dc088d6a42437db6a55f8ee2860b37acc9548e&vid=294ebaf2b8358616561fd5d2c8343a6ab413dedd&prid=web&clid=SAPDAM