BML-210 98 TLC_ANALYSMETOD
Leverantör: ENZO LIFE SCIENCES
Synonyms: CAY10433
BMLGR3300001 BMLGR3300005
ENZOBMLGR3300001EA 1225 SEK
ENZOBMLGR3300001 ENZOBMLGR3300005
BML-210 98 TLC_ANALYSMETOD
BML-210
HDAC inhibitor developed at Enzo Life Sciences. Inhibits HDAC activity in HeLa nuclear extracts (IC50=5-10µM).
  • HDAC inhibitor
Tan solid.
Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml, warm)
Formel: C₂₀H₂₅N₃O₂
MW: 339.43 g/mol
Storage Temperature: Frys
MDL Number: MFCD08062139
CAS nummer: 537034-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4e13ae38c8b295361e899011e590c3638a9940&vid=68f17a50e559d4c67e71d6d33fe5a1a6d2f6cd60&prid=web&clid=SAPDAM