SB-3CT 98 TLC_ANALYSMETOD

Leverantör: ENZO LIFE SCIENCES

Synonyms: 2-(((4-Phenoxyphenyl)sulphonyl)methyl)thiirane

BMLEI3250001 BMLEI3250005
ENZOBMLEI3250001EA 845 SEK
ENZOBMLEI3250001 ENZOBMLEI3250005
SB-3CT 98 TLC_ANALYSMETOD
SB-3CT

Competitive, mechanism-based inhibitor of matrix metalloproteinases 2 and 9 (MMP-2 and MMP-9). It possesses Ki values in the nanomolar range against the gelatinases (14nM for MMP-2; 600nM for MMP-9), but in the micromolar range against other metalloproteinases (206µM for MMP-1, 15µM for MMP-3, 96µM for MMP-7, and 4µM for ADAM17/TACE). This inhibitor has also been used in vitro, in vivo, and in tissue culture. In vivo, it is metabolized to an even more potent gelatinase inhibitor.


  • MMP inhibitor


White solid.


Soluble in DMSO (at least 30mM) or 100% ethanol.

Formel: C₁₅H₁₄O₃S₂
MW: 306.4 g/mol
Storage Temperature: Frys
MDL Number: MFCD09836266
CAS nummer: 292605-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af8f325ba6898ef32cf87084e5d6b29aaa3575bb&vid=cf4639b10cf09620776c00d44abada301d3db3ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR