WH-4-023 ≥99%

Leverantör: ENZO LIFE SCIENCES

ENZCHM1450005 ENZCHM1450010
ENZOENZCHM1450005EA 1475 SEK
ENZOENZCHM1450005 ENZOENZCHM1450010
WH-4-023 ≥99%
WH-4-023

Potent and orally active Lck/Src inhibitor.

Formel: C₃₂H₃₆N₆O₄
MDL Number: MFCD28138072
CAS nummer: 837422-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7a331dcad8444035a99fa94589421a7cabad880&vid=2f66179776e9cc172ddfee31d3778eb4162e70ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR