CCT036477 98 TLC_ANALYSMETOD

Leverantör: ENZO LIFE SCIENCES

BMLWN1140005 BMLWN1140025
ENZOBMLWN1140005EA 1225 SEK
ENZOBMLWN1140005 ENZOBMLWN1140025
CCT036477 98 TLC_ANALYSMETOD
CCT036477

Disrupts Wnt signaling


Inhibits Wnt signaling via blocking transcription at the β-catenin level independent of CBP or p300. Inhibited growth in several cancer cell lines (HT29, HCT116, SW480, and SNU475). CCT036447 was also active in vivo where it blocked development of zebrafish and Xenopus embryos and the expression of Wnt target genes.

MW: 347.8 g/mol
CAS nummer: 305372-78-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecd36f874fe2dcd16784e010730b42a962cebb11&vid=815a2870c738734f7cee7f09bacb53650e52284b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR