H-89 dihydrochloride ≥99%
Leverantör: ENZO LIFE SCIENCES
BMLEI1960005
BMLEI1960025
ENZOBMLEI1960005EA
1850
SEK
ENZOBMLEI1960005
ENZOBMLEI1960025
H-89 dihydrochloride ≥99%
H-89 dihydrochloride
Protein kinase inhibitor
Cell permeable potent and selective inhibitor of cAMP- and cGMP-dependent protein kinases (PKA, Ki=48nM, and PKG) and protein kinase Cµ (PKCµ). In contrast, most other protein kinase C (PKC) isotypes are much more weakly inhibited. Also inhibits Ca2+/calmodulin-dependent protein kinase II, casein kinase I and myosin light chain kinase. Induces apoptosis. Concentration range for cultured cells 30-100μM: osteoblastic cells, renal proximal tubule cells and Xenopus oocytes.
Formel:
C₂₀H₂₀BrN₃O₂S·2HCl |
CAS nummer:
127243-85-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...