NS 1738 ≥99%
Leverantör: ENZO LIFE SCIENCES
Synonyms: 1-(5-chloro-2-hydroxyphenyl)-3-(2-chloro-5-(trifluoromethyl)phenyl)urea, α7 nAChR positive allosteric modulator
ENZCHM1170010 ENZCHM1170050
ENZOENZCHM1170010EA 2000 SEK
ENZOENZCHM1170010 ENZOENZCHM1170050
NS 1738 ≥99%
NS 1738
A selective positive allosteric modulator selective for α7 nicotinic acetylcholine receptors (α7 nAChR).
NS 1738 exhibits cognitive enhancing effects in vivo.
Soluble in DMSO or ethanol.
Formel: C₁₄H₉Cl₂F₃N₂O₂
MW: 365.1 g/mol
Storage Temperature: Frys, ultralågtemperatur (–80°C)
MDL Number: MFCD11100189
CAS nummer: 501684-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8372e8898c3aaa350d086e50b0f3aa6a5c6bd8e8&vid=cb45d68bbb28809812ab68f1ee42cbe41464cbcc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Purity ≥99% (HPLC)