BX-795 ≥97%
Leverantör: ENZO LIFE SCIENCES
Synonyms:
BX795, N-(3-((5-Iodo-4-((3-(thiophene-2-carboxamido)propyl)amino)pyrimidin-2-yl)amino)phenyl)pyrrolidine-1-carboxamide
Inhibitor of PDK1, TBK1, IKK-ε, IRF3
BX795 was initially developed as a PDK1 inhibitor (direct competitive inhibitor IC50 = 11nM for PDK1) and has been shown to be a potent and relatively specific inhibitor of TBK1 and IKK-ε. It blocks the phosphorylation, nuclear translocation, and transcriptional activity of interferon regulatory factor 3. BX795 also is a potent inhibitor of cell growth of multiple cancer cell lines with IC50 values ranging from submicromolar amounts (0.368 µM to greater than 450 µM).
Formel:
C₂₃H₂₆IN₇O₂S MW: 591.5 g/mol |
MDL Number:
MFCD12546134 CAS nummer: 702675-74-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...