E-4031 Dihydrochloride ≥98%
Leverantör: ENZO LIFE SCIENCES
BMLKC1580005
BMLKC1580025
ENZOBMLKC1580005EA
2625
SEK
ENZOBMLKC1580005
ENZOBMLKC1580025
E-4031 Dihydrochloride ≥98%
E-4031 Dihydrochloride
K⁺ channel blocker.
Potent and selective blocker of the inactivating inward rectifying potassium current IKR (HERG channel). Class III antiarrythmic agent with differential atrial vs ventricular activity.
Hygroscopic and soluble in water (100 mg/ml) or DMSO.
Formel:
C₂₁H₂₇N₃O₃S·2HCl·2H₂O MW: 510.5 g/mol Storage Temperature: Omgivande |
CAS nummer:
113558-89-7 |
|
Specifikation testresultat
Purity | ≥98% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...