Astemizole ≥99%, off-white powder

Leverantör: ENZO LIFE SCIENCES
Varning

BMLNH1010010 BMLNH1010050
ENZOBMLNH1010010EA 965 SEK
ENZOBMLNH1010010 ENZOBMLNH1010050
Astemizole ≥99%, off-white powder
Astemizole

Histamine receptor ligand


Selective histamine H1 receptor antagonist.


Soluble in chloroform (clear faint yellow solution at 100 mg/ml).

Formel: C₂₈H₃₁FN₄O
Storage Temperature: Omgivande
MDL Number: MFCD00153919
CAS nummer: 68844-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23a9f7c589b6c3e44e5540dab642079288757003&vid=089bc4efa3c57dd52ef0709087f46e55b82451a6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity ≥99% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR