Cyclopamine ≥98%

Leverantör: ENZO LIFE SCIENCES

BMLGR3340001 BMLGR3340005
ENZOBMLGR3340001EA 1850 SEK
ENZOBMLGR3340001 ENZOBMLGR3340005
Cyclopamine ≥98%
Cyclopamine

Blocks Hedgehog signaling


Cyclopamine is a naturally occuring steroidal alkaloid that causes cyclopia by blocking sonic hedgehog signaling. It specifically inhibits the hedgehog (Hh) pathway by interaction with the Hh signaling protein smoothened. Cyclopamine blocks the oncogenic effects of mutations of Ptch in fibroblasts and inhibits the growth of cells lacking Ptch function. It arrested cell cycle at G0/G1 and induced apoptosis in several small-cell lung cancer cell lines at 5 µM. Cyclopamine also induces expression of the transcription factor pdx-1, which stimulates insulin secretion in differentiation protocols for mouse embryonic stem cells

Formel: C₂₇H₄₁NO₂
MDL Number: MFCD09878269
CAS nummer: 4449-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf0ac48011fab65f5a3729a3769627e1deb28591&vid=b107c25e23a0c92ae54b07560089f83f2dcda4e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR