Ono-RS-082 98 TLC_ANALYSMETOD

Leverantör: ENZO LIFE SCIENCES

BMLST3200020 BMLST3200100
ENZOBMLST3200020EA 990 SEK
ENZOBMLST3200020 ENZOBMLST3200100
Ono-RS-082 98 TLC_ANALYSMETOD
Ono-RS-082

Phospholipase A2 inhibitor (IC50=7 µM for purified guinea pig lung PLA2). Inhibits epinephrine-stimulated thromboxane production in human platelets (86% at 3.5µM). Also blocks insulin secretion in pancreatic islets in response to glucose. Disrupts membrane tubule-mediated reassembly and maintenance of the Golgi complex.


  • Phospholipase inhibitor


Off-white to yellow solid.


Soluble in DMSO (25mg/ml, warm) or 100% ethanol (10mg/ml, warm).

Formel: C₂₁H₂₂ClNO₃
MW: 371.861 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00210755
CAS nummer: 99754-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ee3afaeaccc5027d7cb9976f971d008df86ecc8&vid=9d5003154d9fa9490fb85e789c48ed2d2b22b80c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR