Conduritol B epoxide 96 TLC_ANALYSMETOD

Leverantör: ENZO LIFE SCIENCES

Synonyms: rel-(1R,2R,3S,4S,5R,6S)-7-Oxabicyclo[4.1.0]heptane-2,3,4,5-tetraol

BMLS1040005 BMLS1040025
ENZOBMLS1040005EA 760 SEK
ENZOBMLS1040005 ENZOBMLS1040025
Conduritol B epoxide 96 TLC_ANALYSMETOD
Conduritol B epoxide

A potent, irreversible inhibitor of various plant β-glucosidases and mammalian glucocerebrosidase. Has been employed in in vitro and animal models for Gaucher’s disease. It raises intracellular levels of glucosylceramide by inhibiting glucosylceramidase.


  • Glycosidase inhibitor


White solid.


Soluble in water (25mg/ml) or in DMSO.

Formel: C₆H₁₀O₅
MW: 162.141 g/mol
Storage Temperature: Frys
MDL Number: MFCD00077326
CAS nummer: 6090-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR