monastrol ≥98%
Leverantör: ENZO LIFE SCIENCES
Synonyms: 4-(3-Hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-4H-pyrimidin-5-carboxylic Acid Ethyl Ester, 1,2,3,4-Tetrahydro-4-(3-hydroxyphenyl)-6-methyl-2-thioxo-5-pyrimidinecarboxylic acid, ethyl ester, Ethyl 4-(3-hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-4H-pyrimidin-5-carboxylate, Ethyl 4-(3-hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, Eg5 Inhibitor I
BMLGR3220005 BMLGR3220025
ENZOBMLGR3220005EA 2000 SEK
ENZOBMLGR3220005 ENZOBMLGR3220025
monastrol ≥98%
Monastrol
Cell permeable non-tubulin-interacting mitosis inhibitor. Blocks mitosis (IC50=14µM) by binding to the mitotic kinesin Eg5.
  • Kinesin inhibitor
White to light yellow solid.
Soluble in DMSO (20mg/ml) or 100% ethanol (4mg/ml).
Formel: C₁₄H₁₆N₂O₃S
MW: 292.353 g/mol
Storage Temperature: Frys
MDL Number: MFCD00813077
CAS nummer: 329689-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4859fddc7985a67ca50ceaea49b9a384cfcf104d&vid=86466f37e275695802e2d24d97ff9a88e30ffc6b&prid=web&clid=SAPDAM