Cerulenin ≥98%
Leverantör: ENZO LIFE SCIENCES
BMLG2370005
BMLG2370025
ENZOBMLG2370005EA
1850
SEK
ENZOBMLG2370005
ENZOBMLG2370025
Cerulenin ≥98%
Cerulenin
Fatty acid synthase inhibitor
Antibiotic. Fatty acid synthase (FAS) inhibitor reported to bind in equimolar ratio to β-keto-acyl-ACP synthase (KAS; IC50=1.5µM), thus inhibiting protein acylation at concentrations of 45-134 µM. Produces metabolic effects similar to the effects of leptin, but through mechanisms that are independent of, or down-stream from, both leptin and melanocortin receptors. Apoptosis inducer. Inhibitor of bacterial fatty acid synthesis (inhibits FabH, FabB and FabF condensation enzymes).
Formel:
C₁₂H₁₇NO₃ |
MDL Number:
MFCD00077686 CAS nummer: 17397-89-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...