kainic acid ≥98%, white to off-white solid
Leverantör: ENZO LIFE SCIENCES AG
Synonyms: (2S,3S,4S)-3-(Carboxymethyl)-4-(prop-1-en-2-yl)pyrrolidine-2-carboxylic acid
BMLEA1230005 BMLEA1230025
ENZOBMLEA1230005EA 965 SEK
ENZOBMLEA1230005 ENZOBMLEA1230025
kainic acid ≥98%, white to off-white solid
kainic acid
Selective agonist at kainate receptors.
Soluble in water (25 mg/ml).
Formel: C₁₀H₁₅NO₄
MW: 213,233 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00077806
CAS nummer: 487-79-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc50b335d9ca52ba9c488755b54dd9e6d29df449&vid=8d74394820efe0c9e3d85e351f801565bd8c267c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Purity ≥98% (HPLC)
Identity Determined by H-NMR