Paxilline 97 HPLC_ANALYSMETOD

Leverantör: ENZO LIFE SCIENCES

BMLKC1550005 BMLKC1550025
ENZOBMLKC1550005EA 1850 SEK
ENZOBMLKC1550005 ENZOBMLKC1550025
Paxilline 97 HPLC_ANALYSMETOD
Paxilline

K+ channel blocker


Paxilline is an indole alkaloid isolated from Penicillium paxilli. It is a selective and reversible blocker of the smooth muscle high conductance Ca2+-activated K+ (maxi-K) channel (70% block at 10 nM). In neutrophils, Paxilline blockage of BKCa channels decreased the alkalinization of phagocytic vacuoles and inhibited the microbicidal process. Inhibitor of the cerebellar inositol 1,4,5-triphosphate (InsP(3)) receptor. 

Formel: C₂₇H₃₃NO₄
MW: 435.564 g/mol
MDL Number: MFCD00083464
CAS nummer: 57186-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43f309ef5eff1a14c98901fd1f5086274540077c&vid=92009c6e293287fe6334c7e8cb48cb97202ce9cd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR