Wortmannin ≥98%
Leverantör: ENZO LIFE SCIENCES
Synonyms:
(1S,6bR,9aS,11R,11bR)-1-(Methoxymethyl)-9a,11b-dimethyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3H-furo[4,3,2-de]indeno[4,5-h]isochromen-11-yl acetate
PI3 kinase inhibitor
Potent and selective inhibitor of phosphatidyl-inositol 3-kinase. Active in purified preparations and cytosolic fractions(IC50 = 5nM) and is highly cell permeable. Inhibits fMLP induced PIP3 and superoxide anion production (IC50 = 50nM) in guinea pig neutrophils and blocks the metabolic effects of insulin in isolated rat adipocytes without affecting the insulin receptor tyrosine kinase activity. Covalently binds to PI 3-kinase and is selective, inhibiting other kinases such as PI 4-kinase and myosin light chain kinase at concentrations 100-fold higher than that required for inhibition of PI 3-kinase. The half-life of wortmannin in culture media is quite short (8-13 minutes), care should be taken to account for this decomposition when planning experiments. Markedly potentiates the LPS-induced nitric oxide (NO) production from macrophages. Induces in vivo Alzheimer-like hyperphosphorylation in tau.
Formel:
C₂₃H₂₄O₈ |
MDL Number:
MFCD00133927 CAS nummer: 19545-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...