Mifepristone ≥99%
Leverantör: ENZO LIFE SCIENCES
Fara
BMLS5100025 BMLS5100100
ENZOBMLS5100025EA 1225 SEK
ENZOBMLS5100025 ENZOBMLS5100100
Mifepristone ≥99%
Mifepristone
Progesterone receptor ligand.
Progesterone receptor antagonist.
Soluble in methanol or DMSO.
Formel: C₂₉H₃₅NO₂
Storage Temperature: Frys
CAS nummer: 84371-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=716871e6caa42d45499b47346f5d2417dc14885a&vid=b121b4e51ffa2524223fc91238aa9b18cfcc68cb&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Purity ≥99% (Assay)