NG-Hydroxy-L-arginine monoacetate ≥98%
Leverantör: ENZO LIFE SCIENCES
ALX106004M005 ALX106004M025
ENZOALX106004M005EA 1500 SEK
ENZOALX106004M005 ENZOALX106004M025
NG-Hydroxy-L-arginine monoacetate ≥98%
NG-Hydroxy-L-arginine monoacetate
Intermediate in NO biosynthesis
Intermediate in the biosynthesis of nitric oxide from L-arginine by nitric oxide synthases (NOS). Inhibitor of arginases.
Storage Temperature: Omgivande
CAS nummer: 53598-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f9673b4cc1a7a40ba97b1b986b592300e4928cc&vid=938e0a06d5c1d27579464965c6b2776e45d28bf1&prid=web&clid=SAPDAM